Assay Details
Target Gene Details
Entrez Gene ID: | 56640 |
Gene Name: | kallikrein related-peptidase 4 (prostase, enamel matrix, prostate) |
Gene Aliases: |
ESMP1, KLK-L1, PSTS, Prss17 |
Location: |
Chr.7:43881172-43885804 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk4 | NM_019928.1 | NP_064312.1 | ||
| AF019979.1 | AAC98894.1 | |||
| AK087426.1 | ||||
| BC100716.1 | AAI00717.1 | |||
| BC100717.1 | AAI00718.1 | |||
| BC100718.1 | AAI00719.1 | |||
| BC100719.1 | AAI00720.1 |
More Information
Set Membership: |
|

Genomic Map