Assay Details
Target Gene Details
Entrez Gene ID: | 15377 |
Gene Name: | forkhead box A3 |
Gene Aliases: |
Hnf-3g, Hnf3g, Tcf-3g, Tcf3g |
Location: |
Chr.7:19013283-19023539 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxa3 | NM_008260.2 | 1 | 101 | NP_032286.1 |
| AK019022.1 | 1 | 108 | ||
| BC037083.1 | 1 | 104 | AAH37083.1 | |
| X74938.1 | 1 | 101 | CAA52892.1 |
More Information
Set Membership: |
|

Genomic Map