Assay Details
Target Gene Details
Entrez Gene ID: | 67905 |
Gene Name: | protein phosphatase 1M |
Gene Aliases: |
2810423O19Rik, AW610647, C77250, PP2Ceta |
Location: |
Chr.9:106194953-106199233 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1m | NM_026447.4 | NP_080723.3 | ||
| NM_198931.3 | NP_945149.2 | |||
| AB474373.1 | ||||
| AK013149.1 | BAB28679.2 | |||
| AK017245.1 | BAB30649.1 | |||
| AK087999.1 | BAC40085.1 | |||
| AY332616.1 | AAR01612.1 |
More Information
Set Membership: |
|

Genomic Map