Assay Details
Target Gene Details
Entrez Gene ID: | 16785 |
Gene Name: | ribosomal protein SA |
Gene Aliases: |
67kDa, 67lr, AL022858, Lamr, Lamr1, Lamrl1, MLR, P40, P40-3, P40-8 |
Location: |
Chr.9:120127766-120132369 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpsa | NM_011029.4 | NP_035159.3 | ||
| AF140348.1 | AAD26866.1 | |||
| AK010423.1 | BAB26926.1 | |||
| AK010985.1 | BAB27306.1 | |||
| AK011041.1 | BAB27353.1 | |||
| AK011043.1 | BAB27355.1 | |||
| AK082935.1 | BAC38701.1 | |||
| AK088954.1 | BAC40671.1 | |||
| AK134224.1 | BAE22057.1 | |||
| AK135488.1 | BAE22550.1 | |||
| AK145713.1 | ||||
| AK151123.1 | ||||
| AK160551.1 | BAE35867.1 | |||
| AK160625.1 | BAE35924.1 | |||
| AK165219.1 | BAE38083.1 | |||
| AK166697.1 | BAE38953.1 | |||
| AK166874.1 | BAE39085.1 | |||
| AK167132.1 | BAE39278.1 | |||
| BC003829.1 | AAH03829.1 | |||
| BC037195.1 | AAH37195.1 | |||
| BC055886.1 | AAH55886.1 | |||
| BC081461.1 | AAH81461.1 | |||
| BC084677.1 | AAH84677.1 | |||
| BC092041.1 | AAH92041.1 | |||
| BC094902.1 | AAH94902.1 | |||
| BC099601.1 | AAH99601.1 | |||
| BC110285.1 | AAI10286.1 | |||
| EU554633.1 | ||||
| J02870.1 | AAA39413.1 | |||
| X06406.1 | CAA29696.1 |
More Information
Set Membership: |
|

Genomic Map