Assay Details
Target Gene Details
Entrez Gene ID: | 76964 |
Gene Name: | RIKEN cDNA 2610028H24 gene |
Gene Aliases: |
ORF67 |
Location: |
Chr.10:76449023-76461218 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610028H24Rik | NM_029816.2 | 4 | 432 | NP_084092.2 |
| AK011591.1 | 4 | 388 | BAB27720.1 | |
| AK161552.1 | 4 | 242 | BAE36458.1 | |
| BC051454.1 | 4 | 369 | AAH51454.1 | |
| BQ714071.1 | 4 | 451 |

Genomic Map