Assay Details
Target Gene Details
Entrez Gene ID: | 216001 |
Gene Name: | mitochondrial calcium uptake 1 |
Gene Aliases: |
C730016L05Rik, Calc, Cbara1 |
Location: |
Chr.10:59702482-59864134 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Micu1 | NM_001291442.1 | NP_001278371.1 | ||
| NM_001291443.1 | NP_001278372.1 | |||
| NM_144822.3 | NP_659071.1 | |||
| AK029884.1 | 1 | 87 | ||
| AK046049.1 | BAC32581.1 | |||
| AK049521.1 | BAC33792.1 | |||
| AK050113.1 | BAC34073.1 | |||
| AK051470.1 | ||||
| AK052485.1 | BAC35013.1 | |||
| AK077605.1 | BAC36895.1 | |||
| AK083520.1 | ||||
| AK085553.1 | ||||
| BC018320.1 | AAH18320.1 | |||
| BC023022.1 | AAH23022.1 | |||
| BC026566.1 | AAH26566.1 |
More Information
Set Membership: |
|

Genomic Map