Assay Details
Target Gene Details
Entrez Gene ID: | 13198 |
Gene Name: | DNA-damage inducible transcript 3 |
Gene Aliases: |
CHOP-10, CHOP10, chop, gadd153 |
Location: |
Chr.10:127290793-127296288 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddit3 | NM_001290183.1 | NP_001277112.1 | ||
| NM_007837.4 | NP_031863.3 | |||
| AA929408.1 | ||||
| AK028120.1 | BAE20435.1 | |||
| BC013718.1 | AAH13718.1 | |||
| BY121546.1 | ||||
| X67083.1 | CAA47465.1 |

Genomic Map