Assay Details
Target Gene Details
Entrez Gene ID: | 216443 |
Gene Name: | methionine-tRNA synthetase |
Gene Aliases: |
Metrs, Mtrns |
Location: |
Chr.10:127296221-127311786 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mars | NM_001003913.2 | NP_001003913.1 | ||
| NM_001171582.1 | NP_001165053.1 | |||
| AK178137.1 | ||||
| AK180799.1 | ||||
| AK181384.1 | ||||
| AK184017.1 | ||||
| AK186295.1 | ||||
| AK189447.1 | ||||
| AK193348.1 | ||||
| AK211562.1 | ||||
| AK213658.1 | ||||
| AK214150.1 | ||||
| BC079643.1 | AAH79643.1 |
More Information
Set Membership: |
|

Genomic Map