Assay Details
Target Gene Details
Entrez Gene ID: | 13867 |
Gene Name: | v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) |
Gene Aliases: |
C76256, Erbb-3, Erbb3r, Her3 |
Location: |
Chr.10:128569368-128589501 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erbb3 | NM_010153.1 | NP_034283.1 | ||
| AY686636.1 | AAT95433.1 |
More Information
Set Membership: |
|

Genomic Map