Assay Details
Target Gene Details
Entrez Gene ID: | 237313 |
Gene Name: | interleukin 20 receptor, alpha |
Gene Aliases: |
E230031K19 |
Location: |
Chr.10:19711673-19760053 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il20ra | NM_172786.2 | 6 | 851 | NP_766374.1 |
| AK054215.1 | 6 | 852 | BAC35695.1 | |
| BC056628.1 | 6 | 835 | AAH56628.1 | |
| EF592560.1 |
More Information
Set Membership: |
|

Genomic Map