Assay Details
Target Gene Details
Entrez Gene ID: | 78670 |
Gene Name: | pleckstrin homology domain containing, family J member 1 |
Gene Aliases: |
9530063M10Rik |
Location: |
Chr.10:80796099-80798626 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhj1 | NM_023900.2 | NP_076389.2 | ||
| AK020621.1 | BAB32152.1 | |||
| AK051734.1 | BAC34743.1 | |||
| AK088567.1 | ||||
| AK150144.1 | BAE29339.1 | |||
| AK178993.1 | ||||
| AK219000.1 | ||||
| BC005565.1 | AAH05565.1 | |||
| BE951415.1 |
More Information
Set Membership: |
|

Genomic Map