Assay Details
Target Gene Details
Entrez Gene ID: | 237353 |
Gene Name: | SH3 domain containing ring finger 3 |
Gene Aliases: |
4831416G18Rik, Igfbp5-ip, Sh3md4, Sh3rf31 |
Location: |
Chr.10:58812372-59138916 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3rf3 | NM_172788.3 | NP_766376.2 | ||
| AK029202.1 | BAC26346.1 | |||
| AK132983.1 | BAE21454.1 | |||
| BC057304.1 | AAH57304.1 |

Genomic Map