Assay Details
Target Gene Details
Entrez Gene ID: | 103098 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 15 |
Gene Aliases: |
AA536730, AI326450, AI326451, v7-3 |
Location: |
Chr.10:103367458-103419379 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a15 | NM_001252330.1 | NP_001239259.1 | ||
| NM_175328.3 | NP_780537.1 | |||
| AK036136.1 | BAC29315.1 | |||
| AK082286.1 | 1 | 1594 | ||
| AY149280.1 | AAN75437.1 | |||
| AY149281.1 | AAN75438.1 | |||
| BC076593.1 | AAH76593.1 |
More Information
Set Membership: |
|

Genomic Map