Assay Details
Target Gene Details
Entrez Gene ID: | 76408 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 3 |
Gene Aliases: |
1700019L09Rik, ABC31, MLP2, MOAT-D, MRP3 |
Location: |
Chr.11:94343295-94393022 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcc3 | NM_029600.3 | 12 | 1638 | NP_083876.3 |
| AF534127.1 | 12 | 1638 | AAQ10530.1 | |
| AF534128.1 | 12 | 1638 | AAQ10531.1 | |
| AF534129.1 | 12 | 1638 | AAQ10532.1 | |
| AK149551.1 | 12 | 1638 | BAE28953.1 | |
| AK155664.1 | 12 | 1613 | BAE33375.1 | |
| AK156615.1 | 12 | 1868 | ||
| AK172420.1 | 12 | 1868 | ||
| AY795569.1 | 12 | 1638 | ||
| AY841885.1 | 12 | 1619 | AAX39010.1 | |
| BC048825.1 | 12 | 1570 | AAH48825.1 | |
| BC150788.1 | 12 | 1629 |
More Information
Set Membership: |
|

Genomic Map