Assay Details
Target Gene Details
Entrez Gene ID: | 216725 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 2 |
Gene Aliases: |
ADAM-TS2, ADAMTS-2, ADAMTS-3, PCINP, hPCPNI, mKIAA4060, pNPI |
Location: |
Chr.11:50602085-50807573 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts2 | NM_001277305.1 | 6 | 1136 | NP_001264234.1 |
| NM_175643.3 | 6 | 1136 | NP_783574.1 | |
| AK220464.1 | 5 | 878 | BAD90491.1 | |
| BC046456.1 | 6 | 1136 | AAH46456.1 |
More Information
Set Membership: |
|

Genomic Map