Assay Details
Target Gene Details
Entrez Gene ID: | 11818 |
Gene Name: | apolipoprotein H |
Gene Aliases: |
B2GPI, beta-2-GPI, beta2-GPI |
Location: |
Chr.11:108395297-108414396 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoh | NM_013475.4 | NP_038503.4 | ||
| AK010926.1 | BAB27271.1 | |||
| BC019811.1 | AAH19811.1 | |||
| BC053338.1 | AAH53338.1 | |||
| BY143138.1 | ||||
| D10056.1 | BAA00945.1 | |||
| S70439.1 | AAB30789.1 | |||
| Y11356.1 | CAA72190.1 |

Genomic Map