Assay Details
Target Gene Details
Entrez Gene ID: | 16480 |
Gene Name: | junction plakoglobin |
Gene Aliases: |
Ctnng, D930025P04Rik, PG |
Location: |
Chr.11:100368856-100397790 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jup | NM_010593.2 | 7 | 1274 | NP_034723.1 |
| AK145895.1 | 7 | 1259 | ||
| AK170934.1 | 7 | 1247 | BAE42126.1 | |
| BC040757.1 | 7 | 1223 | AAH40757.1 | |
| BC094461.1 | 7 | 1227 | AAH94461.1 | |
| M90365.1 | 5 | 739 | AAB02885.1 |

Genomic Map