Assay Details
Target Gene Details
Entrez Gene ID: | 72055 |
Gene Name: | solute carrier family 38, member 10 |
Gene Aliases: |
1810073N04Rik |
Location: |
Chr.11:120103951-120151351 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc38a10 | NM_001164798.1 | NP_001158270.1 | ||
| NM_001164799.1 | NP_001158271.1 | |||
| NM_001164800.1 | NP_001158272.1 | |||
| NM_001164801.1 | NP_001158273.1 | |||
| NM_001164802.1 | NP_001158274.1 | |||
| NM_024249.5 | NP_077211.4 | |||
| AK007977.1 | BAB25384.1 | |||
| AK032245.1 | BAC27777.1 | |||
| AK090050.1 | BAC41066.1 | |||
| AK159948.1 | BAE35504.1 | |||
| AK165672.1 | ||||
| AK182610.1 | ||||
| AK190361.1 | ||||
| AK192511.1 | ||||
| AK192970.1 | ||||
| AK208631.1 | ||||
| BC082300.1 | AAH82300.1 | |||
| BC088811.1 | AAH88811.1 |
More Information
Set Membership: |
|

Genomic Map