Assay Details
Target Gene Details
Entrez Gene ID: | 19401 |
Gene Name: | retinoic acid receptor, alpha |
Gene Aliases: |
Nr1b1, RAR, RARalpha1 |
Location: |
Chr.11:98927809-98974942 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rara | NM_001176528.1 | NP_001169999.1 | ||
| NM_001177302.1 | NP_001170773.1 | |||
| NM_001177303.1 | NP_001170774.1 | |||
| NM_009024.2 | NP_033050.2 | |||
| AB221572.1 | ||||
| AK042163.1 | BAE20623.1 | |||
| AK153644.1 | BAE32132.1 | |||
| AK154623.1 | BAE32722.1 | |||
| AK170956.1 | ||||
| BC010216.1 | AAH10216.1 | |||
| CT010231.1 | CAJ18439.1 | |||
| S56656.1 | AAB25783.1 |
More Information
Set Membership: |
|

Genomic Map