Assay Details
Target Gene Details
Entrez Gene ID: | 20370 |
Gene Name: | seizure related gene 6 |
Gene Aliases: |
BSRP-C, D11Bhm177e, sez-6 |
Location: |
Chr.11:77930839-77979052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sez6 | NM_001291225.1 | NP_001278154.1 | ||
| NM_021286.4 | NP_067261.2 | |||
| AB206791.1 | BAE44444.1 | |||
| AK147705.1 | ||||
| AK165757.1 | ||||
| AK220513.1 | BAD90303.1 | |||
| BC053011.1 | AAH53011.1 | |||
| BC055345.1 | AAH55345.1 | |||
| D29763.1 | BAA06167.1 | |||
| D64009.1 | BAA10889.1 | |||
| D64010.1 | BAA10890.1 |

Genomic Map