Assay Details
Target Gene Details
Entrez Gene ID: | 20564 |
Gene Name: | slit homolog 3 (Drosophila) |
Gene Aliases: |
Slil2, Slit1, b2b2362.1Clo |
Location: |
Chr.11:35121456-35708507 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slit3 | NM_011412.3 | 25 | 2741 | NP_035542.2 |
| AF144629.1 | 25 | 2682 | AAD44760.1 | |
| AK147292.1 | 25 | 2742 | BAE27826.1 | |
| BC150780.1 | 25 | 2718 |

Genomic Map