Assay Details
Target Gene Details
Entrez Gene ID: | 70834 |
Gene Name: | sperm associated antigen 9 |
Gene Aliases: |
3110018C07Rik, 4733401I23Rik, 4831406C20Rik, AW552012, JLP, JSAP2, JSAP2a, Jip4, Mapk8ip4, syd1 |
Location: |
Chr.11:93996058-94126087 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 24 - Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spag9 | NM_001025428.1 | NP_001020599.1 | ||
| NM_001025429.1 | NP_001020600.1 | |||
| NM_001025430.1 | NP_001020601.1 | |||
| NM_001199203.1 | NP_001186132.1 | |||
| NM_001199204.1 | NP_001186133.1 | |||
| NM_001199205.1 | NP_001186134.1 | |||
| NM_027569.2 | NP_081845.2 | |||
| AB047782.1 | BAD93176.1 | |||
| AB367798.1 | 3 | 474 | ||
| AF327451.1 | AAN61564.1 | |||
| AK049161.1 | BAC33577.1 | |||
| AK147431.1 | BAE27907.1 | |||
| AK147537.1 | BAE27980.1 | |||
| AK172961.1 | BAD32239.1 | |||
| AY823270.1 | AAX19462.1 | |||
| BC060100.1 | AAH60100.1 | |||
| BC060506.1 | AAH60506.1 | |||
| BC094670.1 | AAH94670.1 | |||
| FJ042496.1 |
More Information
Set Membership: |
|

Genomic Map