Assay Details
Target Gene Details
Entrez Gene ID: | 216835 |
Gene Name: | ubiquitin specific peptidase 43 |
Gene Aliases: |
- |
Location: |
Chr.11:67854523-67922153 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp43 | NM_001291049.1 | 14 | 2346 | NP_001277978.1 |
| NM_173754.4 | 14 | 2361 | NP_776115.2 | |
| AK042217.1 | 14 | 2347 | ||
| AK047339.1 | 2 | 334 | BAC33027.1 | |
| AK083271.1 | 14 | 2199 | BAC38837.1 | |
| BC021474.1 | 13 | 1651 | AAH21474.1 |
More Information
Set Membership: |
|

Genomic Map