Assay Details
Target Gene Details
Entrez Gene ID: | 319504 |
Gene Name: | neuronal cell adhesion molecule |
Gene Aliases: |
Bravo, C030017F07Rik, C130076O07Rik, mKIAA0343 |
Location: |
Chr.12:44328832-44605194 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrcam | NM_001146031.1 | NP_001139503.1 | ||
| NM_176930.4 | NP_795904.3 | |||
| AJ543321.1 | CAD65848.1 | |||
| AK045259.1 | BAC32284.1 | |||
| AK048567.1 | BAC33377.1 | |||
| AK087693.1 | ||||
| AK122252.1 | BAC65534.1 | |||
| AK178439.1 | ||||
| AK187679.1 | ||||
| AK214713.1 | ||||
| AK218057.1 |
More Information
Set Membership: |
|

Genomic Map