Assay Details
Target Gene Details
Entrez Gene ID: | 13665 |
Gene Name: | eukaryotic translation initiation factor 2, subunit 1 alpha |
Gene Aliases: |
0910001O23Rik, 2410026C18Rik, 35kDa, Eif2a, eIF2alpha |
Location: |
Chr.12:78862072-78887010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2s1 | NM_026114.3 | NP_080390.1 | ||
| AK003094.1 | ||||
| AK009934.1 | BAB26593.1 | |||
| AK010592.1 | BAB27049.1 | |||
| AK143499.1 | BAE25400.1 | |||
| AK167763.1 | BAE39796.1 | |||
| BC005463.1 | AAH05463.1 | |||
| BC016448.1 | AAH16448.1 | |||
| BC016497.1 | AAH16497.1 |
More Information
Set Membership: |
|

Genomic Map