Assay Details
Target Gene Details
Entrez Gene ID: | 271005 |
Gene Name: | kelch domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.12:69226917-69284646 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhdc1 | NM_178253.5 | NP_839984.1 | ||
| AK144621.1 | BAE25974.1 | |||
| AY238938.1 | AAO88963.1 | |||
| BC119567.2 | ||||
| BC119568.2 |

Genomic Map