Assay Details
Target Gene Details
Entrez Gene ID: | 217692 |
Gene Name: | signal-induced proliferation-associated 1 like 1 |
Gene Aliases: |
4931426N11Rik, AW213287, Spar, mKIAA0440 |
Location: |
Chr.12:82169378-82451786 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sipa1l1 | NM_001167983.1 | NP_001161455.1 | ||
| NM_172579.3 | NP_766167.2 | |||
| AK029889.1 | BAC26660.1 | |||
| AK051853.1 | ||||
| AK122284.1 | BAC65566.1 | |||
| AK147591.1 | ||||
| AK171876.1 | BAE42714.1 | |||
| BC058681.1 | AAH58681.1 | |||
| BC095958.1 | AAH95958.1 |
More Information
Set Membership: |
|

Genomic Map