Assay Details
Target Gene Details
Entrez Gene ID: | 214579 |
Gene Name: | aldhehyde dehydrogenase family 5, subfamily A1 |
Gene Aliases: |
6330403E24Rik, Ahd-1, Ahd1, D630032B01Rik, OTTMUSG00000000613, SSADH, Ssdh1 |
Location: |
Chr.13:24907579-24937661 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh5a1 | NM_172532.3 | NP_766120.1 | ||
| AK052703.1 | BAC35105.1 | |||
| AK144030.1 | BAE25663.1 | |||
| AK159485.1 | ||||
| AK166313.1 | ||||
| BC138703.1 |
More Information
Set Membership: |
|

Genomic Map