Assay Details
Target Gene Details
Entrez Gene ID: | 50779 |
Gene Name: | regulator of G-protein signaling 6 |
Gene Aliases: |
- |
Location: |
Chr.12:82616176-83158815 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs6 | NM_001282061.1 | NP_001268990.1 | ||
| NM_015812.3 | NP_056627.1 | |||
| AF061933.1 | AAC70011.1 | |||
| AK052527.1 | BAC35026.1 | |||
| AK053087.1 | BAC35262.1 | |||
| AK139323.1 | BAE23955.1 | |||
| BC125029.1 | ||||
| BC125030.1 |

Genomic Map