Assay Details
Target Gene Details
Entrez Gene ID: | 66128 |
Gene Name: | mitochondrial ribosomal protein S36 |
Gene Aliases: |
1110018B13Rik, S36mt |
Location: |
Chr.13:100735940-100744659 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps36 | NM_001190264.1 | NP_001177193.1 | ||
| NM_025369.3 | NP_079645.1 | |||
| AK003765.1 | BAB22983.1 | |||
| AK007512.1 | BAB25081.1 | |||
| AK009050.1 | BAB26047.1 | |||
| BC028499.1 | AAH28499.1 | |||
| BC096576.1 | AAH96576.1 |
More Information
Set Membership: |
|

Genomic Map