Assay Details
Target Gene Details
Entrez Gene ID: | 12522 |
Gene Name: | CD83 antigen |
Gene Aliases: |
- |
Location: |
Chr.13:43785112-43803133 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd83 | NM_001289915.1 | NP_001276844.1 | ||
| NM_009856.3 | NP_033986.1 | |||
| AF001036.1 | AAC83185.1 | |||
| AJ245551.1 | CAB63843.1 | |||
| AK075888.1 | BAC36032.1 | |||
| AK139410.1 | BAE23999.1 | |||
| AK154862.1 | BAE32886.1 | |||
| AK156386.1 | BAE33694.1 | |||
| AK171716.1 | BAE42629.1 | |||
| AK172122.1 | ||||
| AK172396.1 | BAE42984.1 | |||
| AK172449.1 | BAE43011.1 | |||
| AK172466.1 | BAE43020.1 | |||
| AK172515.1 | BAE43044.1 | |||
| BB840485.1 | ||||
| BC107344.1 | AAI07345.1 | |||
| BC107345.1 | AAI07346.1 | |||
| BY336480.1 |

Genomic Map