Assay Details
Target Gene Details
Entrez Gene ID: | 56249 |
Gene Name: | ARP8 actin-related protein 8 |
Gene Aliases: |
5730542K05Rik, ARP8 |
Location: |
Chr.14:29978330-29993221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr8 | NM_027493.3 | NP_081769.2 | ||
| AK017815.1 | BAB30952.1 | |||
| BC027281.1 | AAH27281.1 |
More Information
Set Membership: |
|

Genomic Map