Assay Details
Target Gene Details
Entrez Gene ID: | 22598 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 18 |
Gene Aliases: |
B0AT3, D630001K16Rik, Xt2, Xtrp2 |
Location: |
Chr.13:73661750-73678023 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a18 | NM_001040692.3 | 1 | 57 | NP_001035782.1 |
| NM_001136087.2 | 1 | 57 | NP_001129559.1 | |
| NM_001168643.1 | 1 | 57 | NP_001162114.1 | |
| NM_001168644.1 | 1 | 57 | NP_001162115.1 | |
| NM_001168645.1 | 1 | 57 | NP_001162116.1 | |
| NM_001168646.1 | 1 | 57 | NP_001162117.1 | |
| AF075262.1 | 1 | 35 | AAC27757.1 | |
| AF075263.1 | 1 | 35 | AAC27758.1 | |
| AF075264.1 | 1 | 35 | AAC27759.1 | |
| AF075265.1 | 1 | 35 | AAC27760.1 | |
| AF075266.1 | 1 | 35 | AAC27761.1 | |
| AF075267.1 | 1 | 35 | AAC27762.1 | |
| AK085258.1 | 1 | 54 | ||
| AK085473.1 | 1 | 55 | ||
| AK128907.1 | 1 | 54 | ||
| BC010748.1 | 1 | 33 | AAH10748.1 | |
| CA491609.1 | 1 | 94 |
More Information
Set Membership: |
|

Genomic Map