Assay Details
Target Gene Details
Entrez Gene ID: | 321019 |
Gene Name: | G protein-coupled receptor 183 |
Gene Aliases: |
Ebi2 |
Location: |
Chr.14:121952331-121965193 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr183 | NM_183031.2 | NP_898852.2 | ||
| AK138693.1 | BAE23750.1 |
Target Gene Details
Entrez Gene ID: | 68889 |
Gene Name: | ubiquitin associated domain containing 2 |
Gene Aliases: |
1190008A14Rik, AW554959, Phgdhl1 |
Location: |
Chr.14:121878556-122021035 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubac2 | NM_026861.2 | NP_081137.2 | ||
| AK035384.1 | ||||
| AK049109.1 | BAC33546.1 | |||
| AK158177.1 | BAE34394.1 | |||
| AK199665.1 | ||||
| BC024467.1 | AAH24467.1 |
More Information
Set Membership: |
|

Genomic Map