Assay Details
Target Gene Details
Entrez Gene ID: | 14939 |
Gene Name: | granzyme B |
Gene Aliases: |
AI553453, CCP-1/C11, CCP1, Ctla-1, Ctla1, GZB |
Location: |
Chr.14:56258856-56262260 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gzmb | NM_013542.2 | NP_038570.1 | ||
| AK157856.1 | BAE34234.1 | |||
| AK167770.1 | BAE39803.1 | |||
| BC002085.1 | AAH02085.1 | |||
| CT010272.1 | CAJ18480.1 | |||
| M12302.1 | AAA37383.1 | |||
| X04072.1 | CAA27715.1 |
More Information
Set Membership: |
|

Genomic Map