Assay Details
Target Gene Details
Entrez Gene ID: | 14944 |
Gene Name: | granzyme G |
Gene Aliases: |
Ctla-7, Ctla7 |
Location: |
Chr.14:56156581-56159579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gzmg | NM_010375.2 | NP_034505.1 | ||
| BC099473.1 | AAH99473.1 | |||
| J02872.1 | AAA37731.1 | |||
| M36900.1 | AAA37486.1 | |||
| X14092.1 | CAA32254.1 |
More Information
Set Membership: |
|

Genomic Map