Assay Details
Target Gene Details
Entrez Gene ID: | 76355 |
Gene Name: | TDP-glucose 4,6-dehydratase |
Gene Aliases: |
2610017J16Rik, 2610025M23Rik, AI648925 |
Location: |
Chr.14:118111911-118132765 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgds | NM_029578.3 | NP_083854.3 | ||
| AK011439.1 | ||||
| AK011555.1 | BAB27693.1 | |||
| AK154341.1 | BAE32527.1 | |||
| AK165685.1 | ||||
| AK165955.1 | BAE38483.1 | |||
| BC021419.1 | AAH21419.1 | |||
| BY710811.1 |
More Information
Set Membership: |
|

Genomic Map