Assay Details
Target Gene Details
Entrez Gene ID: | 239463 |
Gene Name: | family with sequence similarity 83, member A |
Gene Aliases: |
- |
Location: |
Chr.15:57985903-58010706 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam83a | NM_173862.2 | NP_776287.2 | ||
| BC030396.1 | AAH30396.1 |

Genomic Map