Assay Details
Target Gene Details
Entrez Gene ID: | 13350 |
Gene Name: | diacylglycerol O-acyltransferase 1 |
Gene Aliases: |
ARAT, C75990, D15Ertd23e, Dgat |
Location: |
Chr.15:76502014-76512020 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgat1 | NM_010046.2 | 2 | 316 | NP_034176.1 |
| AB057816.1 | 2 | 304 | BAC66171.1 | |
| AF078752.1 | 2 | 374 | AAC72917.1 | |
| BC003717.1 | 2 | 316 | AAH03717.1 |
More Information
Set Membership: |
|

Genomic Map