Assay Details
Target Gene Details
Entrez Gene ID: | 105837 |
Gene Name: | Mdm2, transformed 3T3 cell double minute p53 binding protein |
Gene Aliases: |
AI429604, MDM2BP |
Location: |
Chr.15:55557408-55626423 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtbp | NM_001168250.1 | 2 | 370 | NP_001161722.1 |
| NM_134092.3 | 2 | 370 | NP_598853.3 | |
| AJ278508.1 | 2 | 291 | CAC07970.1 | |
| AK080036.1 | 2 | 184 | BAC37810.1 | |
| AK088549.1 | 2 | 184 | BAC40418.1 | |
| AK133855.1 | 2 | 218 | BAE21889.1 | |
| BB615361.1 | 2 | 346 | ||
| BC052197.1 | 2 | 370 | AAH52197.1 |

Genomic Map