Assay Details
Target Gene Details
Entrez Gene ID: | 213012 |
Gene Name: | abhydrolase domain containing 10 |
Gene Aliases: |
- |
Location: |
Chr.16:45729720-45743278 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd10 | NM_001272070.1 | NP_001258999.1 | ||
| NM_172511.4 | NP_766099.3 | |||
| NR_073572.1 | ||||
| AK052670.1 | BAC35091.1 | |||
| AK084998.1 | BAC39335.1 | |||
| AK166025.1 | BAE38528.1 | |||
| AK185534.1 | ||||
| BC027656.1 | AAH27656.1 | |||
| BC058347.1 | AAH58347.1 | |||
| CX237340.1 |
More Information
Set Membership: |
|

Genomic Map