Assay Details
Target Gene Details
Entrez Gene ID: | 71911 |
Gene Name: | 3-hydroxybutyrate dehydrogenase, type 1 |
Gene Aliases: |
2310032J20Rik, AI327223, Bdh |
Location: |
Chr.16:31422297-31458901 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bdh1 | NM_175177.4 | 1 | 71 | NP_780386.3 |
| BC043683.1 | 1 | 74 | AAH43683.1 | |
| BY148101.1 | 1 | 71 |
More Information
Set Membership: |
|

Genomic Map