Assay Details
Target Gene Details
Entrez Gene ID: | 223435 |
Gene Name: | triple functional domain (PTPRF interacting) |
Gene Aliases: |
6720464I07Rik, AA408740, Solo |
Location: |
Chr.15:27730649-28025953 on Build GRCm38 |
Assay Gene Location: | Within Exon 49 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trio | NM_001081302.1 | 49 | 8165 | NP_001074771.1 |
| AK153924.1 | 2 | 255 | BAE32258.1 | |
| BC051169.1 | 8 | 1632 | AAH51169.1 |
More Information
Set Membership: |
|

Genomic Map