Assay Details
Target Gene Details
Entrez Gene ID: | 78618 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 |
Gene Aliases: |
4832442G16, 9530039J15Rik, CNT-B2, Centb2, mKIAA0041 |
Location: |
Chr.16:31092412-31202056 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acap2 | NM_030138.2 | 14 | 1437 | NP_084414.1 |
| AB490692.1 | 15 | 1211 | ||
| AK029343.1 | 14 | 1362 | ||
| AK129041.1 | 14 | 1414 | BAC97851.1 | |
| AK147319.1 | 13 | 1309 | BAE27843.1 | |
| BC046455.1 | AAH46455.1 |

Genomic Map