Assay Details
Target Gene Details
Entrez Gene ID: | 15975 |
Gene Name: | interferon (alpha and beta) receptor 1 |
Gene Aliases: |
CD118, Ifar, Ifnar, Ifrc, Infar |
Location: |
Chr.16:91485087-91510523 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifnar1 | NM_010508.2 | NP_034638.2 | ||
| AK132431.1 | BAE21165.1 | |||
| AK136950.1 | ||||
| AK141630.1 | BAE24775.1 | |||
| AK148246.1 | BAE28435.1 | |||
| AK157555.1 | ||||
| BC043935.1 | AAH43935.1 | |||
| BC052217.1 | AAH52217.1 | |||
| BC052429.1 | AAH52429.1 | |||
| M89641.1 | AAA37890.1 |
More Information
Set Membership: |
|

Genomic Map