Assay Details
Target Gene Details
Entrez Gene ID: | 207683 |
Gene Name: | immunoglobulin superfamily, member 11 |
Gene Aliases: |
1700025L02Rik, BT-IgSF, Igsf13 |
Location: |
Chr.16:38892476-39027159 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf11 | NM_170599.2 | 6 | 941 | NP_733548.2 |
| AB079880.1 | 6 | 778 | BAC07547.1 | |
| AK160146.1 | 6 | 904 | BAE35654.1 | |
| BC057555.1 | 6 | 941 | AAH57555.1 |
More Information
Set Membership: |
|

Genomic Map