Assay Details
Target Gene Details
Entrez Gene ID: | 94112 |
Gene Name: | mediator complex subunit 15 |
Gene Aliases: |
A230074L19Rik, AW536074, Pcqap, mPcqap |
Location: |
Chr.16:17651208-17722947 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med15 | NM_001040683.2 | NP_001035773.1 | ||
| NM_001285884.1 | NP_001272813.1 | |||
| NM_001285886.1 | NP_001272815.1 | |||
| NM_033609.3 | NP_291087.2 | |||
| AF328770.1 | AAK58424.1 | |||
| AK131186.1 | BAD21436.1 | |||
| AK142224.1 | ||||
| AK166522.1 | BAE38826.1 | |||
| AK169904.1 | BAE41448.1 | |||
| BC003221.1 | ||||
| BC049891.1 | AAH49891.1 | |||
| BC054779.1 | AAH54779.1 |

Genomic Map