Assay Details
Target Gene Details
Entrez Gene ID: | 12393 |
Gene Name: | runt related transcription factor 2 |
Gene Aliases: |
AML3, Cbf, Cbfa-1, Cbfa1, LS3, Osf2, PEBP2aA, Pebp2a1, Pebpa2a |
Location: |
Chr.17:44495986-44814831 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Runx2 | NM_001145920.2 | NP_001139392.1 | ||
| NM_001146038.2 | NP_001139510.1 | |||
| NM_001271627.1 | NP_001258556.1 | |||
| NM_001271630.1 | NP_001258559.1 | |||
| NM_001271631.1 | NP_001258560.1 | |||
| NM_009820.5 | NP_033950.2 | |||
| NR_073392.1 | ||||
| NR_073425.1 | ||||
| AF005936.1 | AAB82419.1 | |||
| AF010284.1 | AAB65409.1 | |||
| AF053954.1 | ||||
| AF053955.1 | ||||
| AF053956.1 | AAC78626.1 | |||
| AK089291.1 | ||||
| AK134925.1 | ||||
| AK158925.1 | ||||
| D14636.1 | BAA03485.1 | |||
| D14637.1 | BAA03486.1 | |||
| DQ458792.1 | ABE68713.1 |
More Information
Set Membership: |
|

Genomic Map