Assay Details
Target Gene Details
Entrez Gene ID: | 18701 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class F |
Gene Aliases: |
- |
Location: |
Chr.17:86997258-87025471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigf | NM_008838.1 | NP_032864.1 | ||
| AK132796.1 | BAE21363.1 | |||
| AK171083.1 | BAE42236.1 | |||
| AK218842.1 | ||||
| BC028862.1 | AAH28862.1 | |||
| D50264.1 | BAA08818.1 |
More Information
Set Membership: |
|

Genomic Map