Assay Details
Target Gene Details
Entrez Gene ID: | 18701 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class F |
Gene Aliases: |
- |
Location: |
Chr.17:86997258-87025471 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigf | NM_008838.1 | 2 | 278 | NP_032864.1 |
| AK132796.1 | 2 | 284 | BAE21363.1 | |
| AK171083.1 | 2 | 266 | BAE42236.1 | |
| AK218842.1 | 2 | 190 | ||
| BC028862.1 | 2 | 267 | AAH28862.1 | |
| D50264.1 | 2 | 278 | BAA08818.1 |
More Information
Set Membership: |
|

Genomic Map